4-Chloro-2-fluoro-6-iodoaniline 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17658-03
L17658-06
CAAAAL17658-03EA
59.09
CAD
CAAAAL17658-03
CAAAAL17658-06
4-Chloro-2-fluoro-6-iodoaniline 96%
4-Chloro-2-fluoro-6-iodoaniline
Purity: 96%
CAS No.: 216393-67-8
Molecular Formula: C6H4ClFIN
Formula Weight: 271.46
Melting Point: 39-41°
UN No.: 2811
MDL No.: MFCD01318122
Formula:
C₆H₄ClFIN Melting Pt: 39…41 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318122 CAS Number: 216393-67-8 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...