tert-Butyl (diethylphosphono)acetate 95%

Supplier: Thermo Scientific Chemicals
Warning

L17651-06 L17651-14
CAAAAL17651-03EA 56.56 CAD
CAAAAL17651-03 CAAAAL17651-06 CAAAAL17651-14
tert-Butyl (diethylphosphono)acetate 95%
tert-Butyl (diethylphosphono)acetate

Purity: 95% CAS No.: 27784-76-5 Molecular Formula: C10H21O5P Formula Weight: 252.25 Flash Point: 110°(230°F) Refractive Index: 1.4310 Boiling Point: 100-103°/1.5mm Beilstein Reference: 2050126 MDL No.: MFCD00075414

Formula: C₁₀H₂₁O₅P
Boiling Pt: 100…103 °C/1.5 mm
Density: 1.074
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075414
CAS Number: 27784-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR