5-(tert-Butoxycarbonylamino)-1-pentanol 96%

Supplier: Thermo Scientific Chemicals

Synonyms: N-(5-Hydroxypentyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(5-Hydroxypentyl)carbamate, tert-Butyl (5-hydroxypentyl)carbamate, N-Boc-5-aminopentanol, 5-(Boc-amino)-1-pentanol

L17649-03 L17649-06
CAAAAL17649-03EA 98.28 CAD
CAAAAL17649-03 CAAAAL17649-06
5-(tert-Butoxycarbonylamino)-1-pentanol 96%
5-(tert-Butoxycarbonylamino)-1-pentanol

Purity: 96% CAS No.: 75178-90-4 Molecular Formula: C10H21NO3 Formula Weight: 203.28 Storage Temperature: 2°C to 8°C Flash Point: 109°(228°F) Refractive Index: 1.4560 Beilstein Reference: 3603659 MDL No.: MFCD01862955

Formula: C₁₀H₂₁NO₃
MW: 203.28 g/mol
Density: 1.000
Flash Pt: >109 °C(228 °F)
Storage Temperature: Ambient
MDL Number: MFCD01862955
CAS Number: 75178-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR