1-Isopropenyl-2-benzimidazolidinone ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(Prop-1-en-2-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one, 1-(Prop-1-en-2-yl)-1H-benzo[d]imidazol-2(3H)-one

L17626-03 L17626-06
CAAAAL17626-03EA 60.36 CAD
CAAAAL17626-03 CAAAAL17626-06
1-Isopropenyl-2-benzimidazolidinone ≥98%
1-Isopropenyl-2-benzimidazolidinone

Purity: 98+% CAS No.: 52099-72-6 Molecular Formula: C10H10N2O Formula Weight: 174.20 Melting Point: 121-123° Beilstein Reference: 140587 MDL No.: MFCD00218253

Formula: C₁₀H₁₀N₂O
MW: 174.203 g/mol
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00218253
CAS Number: 52099-72-6
EINECS: 257-661-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR