4-(Ethylthio)benzaldehyde ≥97%
Supplier: Thermo Scientific Chemicals
L17624-03
L17624-06
CAAAAL17624-03EA
102.38
CAD
CAAAAL17624-03
CAAAAL17624-06
4-(Ethylthio)benzaldehyde ≥97%
4-(Ethylthio)benzaldehyde
Purity: 98%
CAS No.: 84211-94-9
Molecular Formula: C9H10OS
Formula Weight: 166.24
Refractive Index: 1.6240
Boiling Point: 151-153°/15mm
MDL No.: MFCD01318145
Formula:
C₉H₁₀OS Boiling Pt: 151…153 °C/15 mm Storage Temperature: Ambient |
MDL Number:
MFCD01318145 CAS Number: 84211-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...