1-Acetylisatin 97%

Supplier: Thermo Scientific Chemicals
Warning

L17597-03 L17597-06 L17597-14
CAAAAL17597-03EA 22.28 CAD
CAAAAL17597-03 CAAAAL17597-06 CAAAAL17597-14
1-Acetylisatin 97%
1-Acetylisatin

Purity: 97% CAS No.: 574-17-4 Molecular Formula: C10H7NO3 Formula Weight: 189.17 Melting Point: 141-143° Beilstein Reference: 113959

Formula: C₁₀H₇NO₃
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00158542
CAS Number: 574-17-4
EINECS: 209-368-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR