Ethyl-5-methoxyindole-2-carboxylate 98%

Supplier: Thermo Scientific Chemicals

L17596-03 L17596-06
CAAAAL17596-03EA 67.78 CAD
CAAAAL17596-03 CAAAAL17596-06
Ethyl-5-methoxyindole-2-carboxylate 98%
Ethyl-5-methoxyindole-2-carboxylate

Purity: 98% CAS No.: 4792-58-9 Molecular Formula: C12H13NO3 Formula Weight: 219.24 Melting Point: 154-157° Beilstein Reference: 178104 MDL No.: MFCD00047163

Formula: C₁₂H₁₃NO₃
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00047163
CAS Number: 4792-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR