Isobutyl-4-hydroxybenzoate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Isobutyl 4-hydroxybenzoate, Isobutylparaben
L17587-06
L17587-18
CAAAAL17587-06EA
52.3
CAD
CAAAAL17587-06
CAAAAL17587-18
Isobutyl-4-hydroxybenzoate 98%
Isobutyl 4-hydroxybenzoate
Purity: 98%
CAS No.: 4247-02-3
Molecular Formula: C11H14O3
Formula Weight: 194.23
Melting Point: 74-78°
Beilstein Reference: 2642305
Formula:
C₁₁H₁₄O₃ Melting Pt: 74…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020167 CAS Number: 4247-02-3 EINECS: 224-208-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...