1-(Phenylsulfonyl)indole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(Phenylsulfonyl)-1H-indole, 1-(Phenylsulphonyl)-1H-indole
L17566-03
L17566-06
CAAAAL17566-03EA
40.45
CAD
CAAAAL17566-03
CAAAAL17566-06
1-(Phenylsulfonyl)indole 98%
1-(Phenylsulfonyl)indole
Purity: 98%
CAS No.: 40899-71-6
Molecular Formula: C14H11NO2S
Formula Weight: 257.31
Melting Point: 77-79°
Boiling Point: 192°/3mm
Beilstein Reference: 1286712
MDL No.: MFCD00134318
Formula:
C₁₄H₁₁NO₂S MW: 257.31 g/mol Boiling Pt: 192 °C/3 mm Melting Pt: 77…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134318 CAS Number: 40899-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...