4,4,5,5-Tetramethyl-1,3,2-dioxaborolane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L17558-06
L17558-14
CAAAAL17558-06EA
87.53
CAD
CAAAAL17558-06
CAAAAL17558-14
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
Purity: 97%
CAS No.: 25015-63-8
Molecular Formula: C6H13BO2
Formula Weight: 127.98
Storage Temperature: 2°C to 8°C
Flash Point: 5°(41°F)
Refractive Index: 1.3970
Boiling Point: 42-43°/50mm
Beilstein Reference: 7802871
UN No.: 3399
MDL No.: MFCD00674030
Formula:
C₆H₁₃BO₂ Boiling Pt: 42…43 °C/50 mm Density: 0.882 Flash Pt: 5 °C(41 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00674030 CAS Number: 25015-63-8 UN: 3399 ADR: "4.3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...