1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(tert-butoxycarbonyl)piperidine-2-carboxylic acid, 1-n-boc-2-piperidinecarboxylic acid, N-BOC-2-Piperidinecarboxylic acid

L17528-03 L17528-06
CAAAAL17528-03EA 34.29 CAD
CAAAAL17528-03 CAAAAL17528-06
1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid 98%
1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid

Purity: 98% CAS No.: 98303-20-9 Molecular Formula: C11H19NO4 Formula Weight: 229.28 Melting Point: 129-131° Beilstein Reference: 480022 MDL No.: MFCD01862877

Formula: C₁₁H₁₉NO₄
MW: 229.276 g/mol
Melting Pt: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD01862877
CAS Number: 98303-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR