1-Boc-piperidine-4-carboxylic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid

L17527-03 L17527-06
CAAAAL17527-03EA 67.31 CAD
CAAAAL17527-03 CAAAAL17527-06
1-Boc-piperidine-4-carboxylic acid 98+%
1-Boc-piperidine-4-carboxylic acid

Purity: 98+% CAS No.: 84358-13-4 Molecular Formula: C11H19NO4 Formula Weight: 229.28 Melting Point: 149-151° MDL No.: MFCD00076999

Formula: C₁₁H₁₉NO₄
MW: 229.3 g/mol
Melting Pt: 149…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00076999
CAS Number: 84358-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR