7-Methylindole-3-carboxyaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17519-03
L17519-06
CAAAAL17519-03EA
135.72
CAD
CAAAAL17519-03
CAAAAL17519-06
CAAAAL17519-MD
7-Methylindole-3-carboxyaldehyde 98%
7-Methylindole-3-carboxyaldehyde
Purity: 98%
CAS No.: 4771-50-0
Molecular Formula: C10H9NO
Formula Weight: 159.19
Storage Temperature: 2°C to 8°C
Melting Point: 209-211°
Beilstein Reference: 122340
MDL No.: MFCD00047170
Formula:
C₁₀H₉NO Melting Pt: 209…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047170 CAS Number: 4771-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...