2-Amino-5-chlorobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17515-03
L17515-06
CAAAAL17515-03EA
121.34
CAD
CAAAAL17515-03
CAAAAL17515-06
2-Amino-5-chlorobenzaldehyde 97%
2-Amino-5-chlorobenzaldehyde
Purity: 97%
CAS No.: 20028-53-9
Molecular Formula: C7H6ClNO
Formula Weight: 155.58
Melting Point: 68-74°
Beilstein Reference: 2716096
Formula:
C₇H₆ClNO MW: 155.58 g/mol Melting Pt: 68…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093936 CAS Number: 20028-53-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...