5-Fluoro-1H-indole-3-carbaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

L17507-03 L17507-06 L17507-MD
CAAAAL17507-03EA 222.78 CAD
CAAAAL17507-03 CAAAAL17507-06 CAAAAL17507-MD
5-Fluoro-1H-indole-3-carbaldehyde 98%
5-Fluoro-1H-indole-3-carbaldehyde

Purity: 98% CAS No.: 2338-71-8 Molecular Formula: C9H6FNO Formula Weight: 163.15 Melting Point: 164° Beilstein Reference: 1449216 MDL No.: MFCD00022719

Formula: C₉H₆FNO
Melting Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00022719
CAS Number: 2338-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR