Ethyl-4-acetyl-5-oxohexanoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 4,4-Diacetylbutyrate, Ethyl 4-acetyl-5-oxohexanoate, 4,4-Diacetylbutyric Acid Ethyl Ester, 4-Acetyl-5-oxohexanoic Acid Ethyl Ester

L17496-06 L17496-14
CAAAAL17496-06EA 24.17 CAD
CAAAAL17496-06 CAAAAL17496-14
Ethyl-4-acetyl-5-oxohexanoate 98%
Ethyl-4-acetyl-5-oxohexanoate

Purity: 98% CAS No.: 2832-10-2 Molecular Formula: C10H16O4 Formula Weight: 200.24 Flash Point: 110°(230°F) Refractive Index: 1.4545 Boiling Point: 153-154°/19mm Beilstein Reference: 1785189 MDL No.: MFCD00009211

Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
Boiling Pt: 153…154 °C/19 mm
Density: 1.067
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009211
CAS Number: 2832-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR