4-(2-Carboxyethyl)phenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17485-03
L17485-06
CAAAAL17485-03EA
173.8
CAD
CAAAAL17485-03
CAAAAL17485-06
4-(2-Carboxyethyl)phenylboronic acid 97%
4-(2-Carboxyethyl)phenylboronic acid
Purity: 97%
CAS No.: 166316-48-9
Molecular Formula: C9H11BO4
Formula Weight: 193.99
Melting Point: 146-148°
Beilstein Reference: 9198524
Formula:
C₉H₁₁BO₄ Melting Pt: 146…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318119 CAS Number: 166316-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...