5-Isopropyl-2-methoxybenzeneboronic acid ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (5-Isopropyl-2-methoxyphenyl)boronic acid

L17460-03 L17460-06
CAAAAL17460-03EA 80.9 CAD
CAAAAL17460-03 CAAAAL17460-06
5-Isopropyl-2-methoxybenzeneboronic acid ≥98%
5-Isopropyl-2-methoxybenzeneboronic acid

Purity: 98+% CAS No.: 216393-63-4 Molecular Formula: C10H15BO3 Formula Weight: 194.04 Melting Point: 85-87° Beilstein Reference: 9193266 MDL No.: MFCD01318154

Formula: C₁₀H₁₅BO₃
MW: 194.04 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD01318154
CAS Number: 216393-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR