Tetrahydropyran-2-methanol 94%

Supplier: Thermo Scientific Chemicals
Warning

L17417-06 L17417-14
CAAAAL17417-06EA 66.83 CAD
CAAAAL17417-06 CAAAAL17417-14
Tetrahydropyran-2-methanol 94%
Tetrahydropyran-2-methanol

Purity: 94% CAS No.: 100-72-1 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 93°(199°F) Refractive Index: 1.4580 Melting Point: -70° Boiling Point: 186-189° Beilstein Reference: 102998 MDL No.: MFCD00006624

Formula: C₆H₁₂O₂
Boiling Pt: 186…189 °C
Melting Pt: < –70 °C
Density: 1.027
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006624
CAS Number: 100-72-1
EINECS: 202-882-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR