2-Hydroxy-2-(trifluoromethyl)propionic acid 94%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L17399-03
L17399-06
CAAAAL17399-03EA
68.1
CAD
CAAAAL17399-03
CAAAAL17399-06
2-Hydroxy-2-(trifluoromethyl)propionic acid 94%
2-Hydroxy-2-(trifluoromethyl)propionic acid
Purity: 94%
CAS No.: 114715-77-4
Molecular Formula: C4H5F3O3
Formula Weight: 158.08
Melting Point: ca 85°
Boiling Point: 189°
Beilstein Reference: 1723691
UN No.: 3261
MDL No.: MFCD00190646
Formula:
C₄H₅F₃O₃ MW: 158.08 g/mol Boiling Pt: 189 °C Melting Pt: ∼ 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190646 CAS Number: 114715-77-4 EINECS: 609-428-8 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...