Ethyl-2-methyl-4,4,4-trifluoroacetoacetate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17335-03
L17335-06
CAAAAL17335-03EA
58.78
CAD
CAAAAL17335-03
CAAAAL17335-06
Ethyl-2-methyl-4,4,4-trifluoroacetoacetate 96%
Ethyl-2-methyl-4,4,4-trifluoroacetoacetate
Purity: 96%
CAS No.: 344-00-3
Molecular Formula: C7H9F3O3
Formula Weight: 198.14
Flash Point: 47°(116°F)
Refractive Index: 1.3700
Boiling Point: 144-146°
Beilstein Reference: 1783620
UN No.: 3272
MDL No.: MFCD00190635
Formula:
C₇H₉F₃O₃ Boiling Pt: 144…146 °C Density: 1.258 Flash Pt: 47 °C(116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190635 CAS Number: 344-00-3 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...