N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine 90%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Chloro-N,N-diethyl-1,1,2-trifluoroethylamine

L17330-06 L17330-14 L17330-22
CAAAAL17330-06EA 76.16 CAD
CAAAAL17330-06 CAAAAL17330-14 CAAAAL17330-22
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine 90%
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine

Purity: 90% CAS No.: 357-83-5 Molecular Formula: C6H11ClF3N Formula Weight: 189.61 Storage Temperature: 2°C to 8°C Boiling Point: 38°/10mm Beilstein Reference: 1098675 UN No.: 3267 MDL No.: MFCD00054672

Formula: C₆H₁₁ClF₃N
MW: 189.61 g/mol
Boiling Pt: 38 °C/10 mm
Density: 1.190
Storage Temperature: Ambient
MDL Number: MFCD00054672
CAS Number: 357-83-5
UN: 3267
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR