10,11-Dihydrocarbamazepine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carboxamide

L17271-MD L17271-MF
CAAAAL17271-MDEA 232.26 CAD
CAAAAL17271-MD CAAAAL17271-MF
10,11-Dihydrocarbamazepine 99%
10,11-Dihydrocarbamazepine

Purity: 99% CAS No.: 3564-73-6 Molecular Formula: C15H14N2O Formula Weight: 238.29 Melting Point: 204-206° Beilstein Reference: 196969

Formula: C₁₅H₁₄N₂O
MW: 238.29 g/mol
Melting Pt: 204…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00005072
CAS Number: 3564-73-6
EINECS: 222-649-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR