1-Ethynylpyrene 96%
Supplier: Thermo Scientific Chemicals
L17227-03
L17227-06
CAAAAL17227-03EA
156.74
CAD
CAAAAL17227-03
CAAAAL17227-06
1-Ethynylpyrene 96%
1-Ethynylpyrene
Purity: 96%
CAS No.: 34993-56-1
Molecular Formula: C18H10
Formula Weight: 226.28
Melting Point: 113-116°
Beilstein Reference: 2365008
MDL No.: MFCD02093933
Formula:
C₁₈H₁₀ Melting Pt: 113…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093933 CAS Number: 34993-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...