3-Ethynyltoluene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Ethynyl-3-methylbenzene, m-Tolylacetylene
L17184-03
L17184-06
CAAAAL17184-03EA
66.36
CAD
CAAAAL17184-03
CAAAAL17184-06
3-Ethynyltoluene 97%
3-Ethynyltoluene
Purity: 97%
CAS No.: 766-82-5
Molecular Formula: C9H8
Formula Weight: 116.16
Flash Point: 50°(122°F)
Refractive Index: 1.5440
Odor: Pungent
Boiling Point: 170-175°
Beilstein Reference: 2038007
UN No.: 3295
MDL No.: MFCD01318174
Formula:
C₉H₈ Boiling Pt: 170…175 °C Density: 0.900 Flash Pt: 50 °C(122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01318174 CAS Number: 766-82-5 UN: 3295 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...