tert-Butyl acrylate 99% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acrylic Acid tert-Butyl Ester
Stabilization:
15 ppm Stabilised with 4-methoxyphenol (MEHQ)
L17169-AE
L17169-AP
L17169-AU
CAAAAL17169-AEEA
58.62
CAD
CAAAAL17169-AE
CAAAAL17169-AP
CAAAAL17169-AU
tert-Butyl acrylate 99% stabilised
tert-Butyl acrylate
Stab. with 15ppm 4-methoxyphenol
Purity: 99%
CAS No.: 1663-39-4
Molecular Formula: C7H12O2
Formula Weight: 128.17
Flash Point: 11°(52°F)
Refractive Index: 1.4110
Melting Point: ca -69°
Boiling Point: 121°
Beilstein Reference: 1742329
UN No.: 3272
MDL No.: MFCD00008809
Formula:
C₇H₁₂O₂ MW: 128.17 g/mol Boiling Pt: 121 °C Melting Pt: ∼ 69 °C Density: 0.883 Flash Pt: 11 °C (52 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008809 CAS Number: 1663-39-4 EINECS: 216-768-7 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...