Ethyl-5-methylindole-2-carboxylate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 5-methylindole-2-carboxylate

L17146-03 L17146-06
CAAAAL17146-03EA 59.25 CAD
CAAAAL17146-03 CAAAAL17146-06
Ethyl-5-methylindole-2-carboxylate 98%
Ethyl-5-methylindole-2-carboxylate

Purity: 99% CAS No.: 16382-15-3 Molecular Formula: C12H13NO2 Formula Weight: 203.24 Melting Point: 161-165° Boiling Point: 236°/4mm Beilstein Reference: 159437 MDL No.: MFCD00022703

Formula: C₁₂H₁₃NO₂
MW: 203.24 g/mol
Boiling Pt: 236 °C/4 mm
Melting Pt: 161…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00022703
CAS Number: 16382-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR