6-Cyanonicotinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17127-03
L17127-MD
CAAAAL17127-03EA
290.72
CAD
CAAAAL17127-03
CAAAAL17127-MD
6-Cyanonicotinic acid 97%
6-Cyanonicotinic acid
Purity: 97%
CAS No.: 70165-31-0
Molecular Formula: C7H4N2O2
Formula Weight: 148.12
Melting Point: 189-194°
Beilstein Reference: 4384027
MDL No.: MFCD01318135
Formula:
C₇H₄N₂O₂ Melting Pt: 189…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318135 CAS Number: 70165-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...