Hydrogen fluoride pyridine complex (∼70% HF)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Olah′s reagent, Pyridinium polybifluoride, Pyridine hydrofluoride, HF-Pyridine, PPHF, Poly(pyridine fluoride), Pyridinium poly(hydrogen fluoride)
L17117-14
L17117-22
CAAAAL17117-14EA
96.54
CAD
CAAAAL17117-14
CAAAAL17117-22
Hydrogen fluoride pyridine complex (∼70% HF)
Hydrogen fluoride pyridine complex
Fieser: 5,538 6,285,473 9,399 12,419 16,286 18,181
CAS No.: 62778-11-4
Storage Temperature: 2°C to 8°C
Boiling Point: 50°/1mm
Beilstein Reference: 4750151
UN No.: 1790
MDL No.: MFCD00012436
Formula:
C₅H₅N·(HF)x Boiling Pt: 50 °C/1 mm Density: 1.100 Storage Temperature: Ambient |
MDL Number:
MFCD00012436 CAS Number: 62778-11-4 UN: 1790 ADR: 8 II Merck Index: 14,6821 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...