1-Phenyl-1,2,3-butanetrione-2-oxime ≥98%

Supplier: Thermo Scientific Chemicals
Danger

L17073-03 L17073-06
CAAAAL17073-03EA 74.42 CAD
CAAAAL17073-03 CAAAAL17073-06
1-Phenyl-1,2,3-butanetrione-2-oxime ≥98%
1-Phenyl-1,2,3-butanetrione-2-oxime

Purity: 98+% CAS No.: 6797-44-0 Molecular Formula: C10H9NO3 Formula Weight: 191.19 Odor: Odorless Melting Point: 128-130° Beilstein Reference: 1946580 MDL No.: MFCD00466576

Formula: C₁₀H₉NO₃
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00466576
CAS Number: 6797-44-0
EINECS: 229-869-6
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR