Bis(4-chlorobutyl)ether 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,4'-Dichlorodibutyl Ether, 4,4'-Dichlorobutyl Ether, 4-Chlorobutyl Ether

L17055-06 L17055-14 L17055-22
CAAAAL17055-06EA 41.87 CAD
CAAAAL17055-06 CAAAAL17055-14 CAAAAL17055-22
Bis(4-chlorobutyl)ether 99%
Bis(4-chlorobutyl)ether

Purity: 99% CAS No.: 6334-96-9 Molecular Formula: C8H16Cl2O Formula Weight: 199.12 Refractive Index: 1.4580 Boiling Point: 129-131°/10mm Beilstein Reference: 1739861 MDL No.: MFCD00019006

Formula: C₈H₁₆Cl₂O
MW: 199.12 g/mol
Boiling Pt: 129…131 °C/10 mm
Density: 1.081
Storage Temperature: Ambient
MDL Number: MFCD00019006
CAS Number: 6334-96-9
EINECS: 228-713-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR