4-Methyl-3-nitrophenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Methyl-3-nitrophenyl)boronic acid, 4-Methyl-3-nitrobenzeneboronic acid

L17052-03 L17052-06
CAAAAL17052-03EA 41.87 CAD
CAAAAL17052-03 CAAAAL17052-06
4-Methyl-3-nitrophenylboronic acid 98%
4-Methyl-3-nitrophenylboronic acid

Purity: 98% CAS No.: 80500-27-2 Molecular Formula: C7H8BNO4 Formula Weight: 180.96 Melting Point: ca 250° Beilstein Reference: 3279018

Formula: C₇H₈BNO₄
MW: 180.96 g/mol
Melting Pt: ∼ 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00191550
CAS Number: 80500-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR