Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate 97%
Supplier: Thermo Scientific Chemicals
L17046-03
L17046-MD
CAAAAL17046-03EA
486.64
CAD
CAAAAL17046-03
CAAAAL17046-MD
Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate 97%
Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate
Purity: 97%
CAS No.: 72022-68-5
Molecular Formula: C22H40N2S10Zn
Formula Weight: 718.60
Melting Point: 202-206°
Beilstein Reference: 4900651
MDL No.: MFCD00623815
Formula:
C₂₂H₄₀N₂S₁₀Zn Melting Pt: 202…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00623815 CAS Number: 72022-68-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...