(1S,2S)-(+)-2-Benzyloxycyclohexylamine ≥99%, ee 99%, ChiPros®

Supplier: Thermo Scientific Chemicals
Danger

L17020-03 L17020-06
CAAAAL17020-03EA 89.59 CAD
CAAAAL17020-03 CAAAAL17020-06
(1S,2S)-(+)-2-Benzyloxycyclohexylamine ≥99%, ee 99%, ChiPros®
(1S,2S)-(+)-2-Benzyloxycyclohexylamine

Purity: 99+% CAS No.: 216394-07-9 Molecular Formula: C13H19NO Formula Weight: 205.30 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5275 Boiling Point: 79-80°/0.08mm Beilstein Reference: 8981794 Optical Rotation: +85°(neat) UN No.: 2735 MDL No.: MFCD01075753

Formula: C₁₃H₁₉NO
MW: 205.30 g/mol
Boiling Pt: 79…80 °C/0.08 mm
Density: 1.02
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01075753
CAS Number: 216394-07-9
EINECS: 606-811-1
UN: 2735
ADR: "8",III

Order Now

Specification Test Results

Optical rotation +85°(neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR