3-Bromo-2-chloropyridine-5-sulfonyl chloride 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Bromo-6-chloropyridine-3-sulphonyl chloride

L17015-03 L17015-06
CAAAAL17015-03EA 94.48 CAD
CAAAAL17015-03 CAAAAL17015-06
3-Bromo-2-chloropyridine-5-sulfonyl chloride 96%
3-Bromo-2-chloropyridine-5-sulfonyl chloride

Purity: 96% CAS No.: 216394-05-7 Molecular Formula: C5H2BrCl2NO2S Formula Weight: 290.95 Melting Point: 65-70° UN No.: 3261 MDL No.: MFCD01318107

Formula: C₅H₂BrCl₂NO₂S
MW: 290.96 g/mol
Melting Pt: 65…70 °C
Storage Temperature: Ambient
MDL Number: MFCD01318107
CAS Number: 216394-05-7
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR