N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(Chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane Bis(tetrafluoroborate), F-TEDA-BF4, 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)

L17003-06 L17003-14
CAAAAL17003-06EA 87.69 CAD
CAAAAL17003-06 CAAAAL17003-14
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥98%
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)

Fieser: 18,100 20,106 21,129


Purity: 98+% CAS No.: 140681-55-6 Molecular Formula: C7H14B2ClF9N2 Formula Weight: 354.26 Melting Point: ca 250° dec. UN No.: 3088 MDL No.: MFCD00142607

Formula: C₇H₁₄B₂ClF₉N₂
MW: 354.26 g/mol
Melting Pt: ∼ 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00142607
CAS Number: 140681-55-6
EINECS: 414-380-4
UN: 3088
ADR: 4.2,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR