2-Methoxy-4-methylbenzenesulfonamide 95%
Supplier: Thermo Scientific Chemicals
L16947-03
L16947-06
CAAAAL16947-03EA
42.34
CAD
CAAAAL16947-03
CAAAAL16947-06
2-Methoxy-4-methylbenzenesulfonamide 95%
2-Methoxy-4-methylbenzenesulfonamide
Purity: 95%
CAS No.: 59554-39-1
Molecular Formula: C8H11NO3S
Formula Weight: 201.25
Melting Point: 168-170°
MDL No.: MFCD01318156
Formula:
C₈H₁₁NO₃S Melting Pt: 168…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318156 CAS Number: 59554-39-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...