3-(Trifluoromethyl)-3-hydroxybutyric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,4,4-Trifluoro-3-hydroxy-3-methylbutanoic acid, 3-Hydroxy-3-(trifluoromethyl)butyric acid

L16873-03 L16873-06
CAAAAL16873-03EA 77.26 CAD
CAAAAL16873-03 CAAAAL16873-06
3-(Trifluoromethyl)-3-hydroxybutyric acid 98%
3-(Trifluoromethyl)-3-hydroxybutyric acid

Purity: 98% CAS No.: 338-03-4 Molecular Formula: C5H7F3O3 Formula Weight: 172.10 Refractive Index: 1.3880 Melting Point: 39-41° Boiling Point: 87-90°/0.5mm UN No.: 3261 MDL No.: MFCD00039531

Formula: C₅H₇F₃O₃
MW: 172.1 g/mol
Boiling Pt: 87…90 °C/0.5 mm
Melting Pt: 39…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00039531
CAS Number: 338-03-4
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR