2,2,3,3,3-Pentafluoropropylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

L16819-03 L16819-06
CAAAAL16819-03EA 164.32 CAD
CAAAAL16819-03 CAAAAL16819-06
2,2,3,3,3-Pentafluoropropylamine 97%
2,2,3,3,3-Pentafluoropropylamine

Purity: 97% CAS No.: 422-03-7 Molecular Formula: CF3CF2CH2NH2 Formula Weight: 149.06 Storage Temperature: 2°C to 8°C Flash Point: 100°(212°F) Refractive Index: 1.2970 Boiling Point: 49-50° Beilstein Reference: 1743135 UN No.: 2735 MDL No.: MFCD00042459

Formula: C₃H₄F₅N
MW: 149.06 g/mol
Boiling Pt: 49…50 °C
Density: 1.400
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042459
CAS Number: 422-03-7
EINECS: 207-010-6
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR