Lithium trifluoroacetate 97%

Supplier: Thermo Scientific Chemicals
Danger

L16801-09 L16801-18
CAAAAL16801-09EA 60.2 CAD
CAAAAL16801-09 CAAAAL16801-18
Lithium trifluoroacetate 97%
Lithium trifluoroacetate

Purity: 97% CAS No.: 2923-17-3 Molecular Formula: LiCO2CF3 Formula Weight: 119.96 UN No.: 2811 MDL No.: MFCD00013216

Formula: LiCO₂CF₃
MW: 119.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013216
CAS Number: 2923-17-3
EINECS: 220-878-0
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR