2-Iodononafluorobutane 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with copper
L16792-06
L16792-14
CAAAAL16792-06EA
80.58
CAD
CAAAAL16792-06
CAAAAL16792-14
2-Iodononafluorobutane 98% stabilized
2-Iodononafluorobutane
Stab. with copper
Purity: 98%
CAS No.: 375-51-9
Molecular Formula: CF3CF2CFICF3
Formula Weight: 345.93
Storage Temperature: 2°C to 8°C
Refractive Index: 1.3305
Boiling Point: 64-67°
Beilstein Reference: 1777547
MDL No.: MFCD00077556
Formula:
C₄F₉I Boiling Pt: 64…67 °C Density: 2.095 Storage Temperature: Ambient |
MDL Number:
MFCD00077556 CAS Number: 375-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...