3,3'-Difluorobiphenyl 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16740-03
L16740-06
CAAAAL16740-03EA
164.32
CAD
CAAAAL16740-03
CAAAAL16740-06
3,3'-Difluorobiphenyl 97%
3,3'-Difluorobiphenyl
Purity: 97%
CAS No.: 396-64-5
Molecular Formula: C12H8F2
Formula Weight: 190.19
Refractive Index: 1.5680
Melting Point: 7-8°
Boiling Point: 130°/14mm
Beilstein Reference: 2046445
UN No.: 3151
MDL No.: MFCD00039216
Formula:
C₁₂H₈F₂ Boiling Pt: 130 °C/14 mm Melting Pt: 7…8 °C Density: 1.210 Storage Temperature: Ambient |
MDL Number:
MFCD00039216 CAS Number: 396-64-5 EINECS: 206-905-9 UN: 3151 ADR: "9",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...