2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Chloro-1H,4H-perfluoro(2-oxabutane)
L16725-06
L16725-14
CAAAAL16725-06EA
105.23
CAD
CAAAAL16725-06
CAAAAL16725-14
2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether 97%
2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether
Purity: 97%
CAS No.: 13838-16-9
Molecular Formula: C3H2ClF5O
Formula Weight: 184.49
Storage Temperature: 2°C to 8°C
Flash Point: None
Refractive Index: 1.3030
Boiling Point: 56-57°
Beilstein Reference: 1737129
MDL No.: MFCD00069095
Formula:
C₃H₂ClF₅O Boiling Pt: 56…57 °C Density: 1.517 Flash Pt: None Storage Temperature: Ambient |
MDL Number:
MFCD00069095 CAS Number: 13838-16-9 EINECS: 237-553-4 Merck Index: 14,3581 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...