Bis(perfluoroisopropyl)ketone 97%

Supplier: Thermo Scientific Chemicals
Warning

L16708-03 L16708-06
CAAAAL16708-03EA 56.72 CAD
CAAAAL16708-03 CAAAAL16708-06
Bis(perfluoroisopropyl)ketone 97%
Bis(perfluoroisopropyl)ketone

Purity: 97% CAS No.: 813-44-5 Molecular Formula: C7F14O Formula Weight: 366.06 Boiling Point: 72-73° Beilstein Reference: 1916532 UN No.: 2810 MDL No.: MFCD00042087

Formula: C₇F₁₄O
Boiling Pt: 72…73 °C
Density: 1.661
Storage Temperature: Ambient
MDL Number: MFCD00042087
CAS Number: 813-44-5
EINECS: 212-384-9
UN: 2810
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR