Dimethylethoxyvinylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16699-14
L16699-22
CAAAAL16699-06EA
78.49
CAD
CAAAAL16699-06
CAAAAL16699-14
CAAAAL16699-22
Dimethylethoxyvinylsilane 97%
Dimethylethoxyvinylsilane
Purity: 97%
CAS No.: 5356-83-2
Molecular Formula: C6H14OSi
Formula Weight: 130.26
Flash Point: -3°(26°F)
Refractive Index: 1.3980
Boiling Point: 99°
Beilstein Reference: 1743477
UN No.: 1993
MDL No.: MFCD00009070
Formula:
C₆H₁₄OSi Boiling Pt: 99 °C Density: 0.790 Flash Pt: -3 °C(26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009070 CAS Number: 5356-83-2 EINECS: 226-341-7 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...