Triethoxyfluorosilane 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16652-06
L16652-14
CAAAAL16652-06EA
257.54
CAD
CAAAAL16652-06
CAAAAL16652-14
Triethoxyfluorosilane 95%
Triethoxyfluorosilane
Purity: 95%
CAS No.: 358-60-1
Molecular Formula: C6H15FO3Si
Formula Weight: 182.27
Flash Point: 19°(66°F)
Refractive Index: 1.3610
Boiling Point: 134-135°
Beilstein Reference: 1700825
UN No.: 2924
MDL No.: MFCD00236719
Formula:
C₆H₁₅FO₃Si Boiling Pt: 134…135 °C Density: 0.900 Flash Pt: 19 °C(66 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00236719 CAS Number: 358-60-1 UN: 2924 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...