1H,1H,2H,2H-Perfluorooctyltrichlorosilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-trichlorosilane
L16606-09
L16606-18
CAAAAL16606-09EA
159.58
CAD
CAAAAL16606-09
CAAAAL16606-18
1H,1H,2H,2H-Perfluorooctyltrichlorosilane 97%
1H,1H,2H,2H-Perfluorooctyltrichlorosilane
Purity: 97%
CAS No.: 78560-45-9
Molecular Formula: CF3(CF2)5(CH2)2SiCl3
Formula Weight: 481.55
Flash Point: 87°(189°F)
Refractive Index: 1.3520
Boiling Point: 192°
Beilstein Reference: 6154526
UN No.: 2987
MDL No.: MFCD00042363
Formula:
C₈H₄Cl₃F₁₃Si Boiling Pt: 192 °C Density: 1.638 Flash Pt: 87 °C(189 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042363 CAS Number: 78560-45-9 EINECS: 278-947-6 UN: 2987 ADR: "8",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...