1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane 97%

Supplier: Thermo Scientific Chemicals
Danger

L16605-06 L16605-14
CAAAAL16605-06EA 57.35 CAD
CAAAAL16605-06 CAAAAL16605-14
1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane 97%
1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane

Purity: 97% CAS No.: 73609-36-6 Molecular Formula: C9H7Cl2F13Si Formula Weight: 461.13 Flash Point: 51°(123°F) Refractive Index: 1.3500 Boiling Point: 189-190° Beilstein Reference: 6153667 UN No.: 2920 MDL No.: MFCD00042360

Formula: C₉H₇Cl₂F₁₃Si
MW: 461.13 g/mol
Boiling Pt: 189…190 °C
Density: 1.550
Flash Pt: 51 °C(123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042360
CAS Number: 73609-36-6
EINECS: 277-551-0
UN: 2920
ADR: "8",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR