Undecafluoro(2-methyl-3-oxahexanoyl)fluoride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16483-06
L16483-14
CAAAAL16483-03EA
107.33
CAD
CAAAAL16483-03
CAAAAL16483-06
CAAAAL16483-14
Undecafluoro(2-methyl-3-oxahexanoyl)fluoride 97%
Undecafluoro(2-methyl-3-oxahexanoyl)fluoride
Purity: 97%
CAS No.: 2062-98-8
Molecular Formula: C6F12O2
Formula Weight: 332.06
Flash Point: None
Refractive Index: 1.3000
Boiling Point: 54-56°
UN No.: 3265
MDL No.: MFCD00054657
Formula:
C₆F₁₂O₂ Boiling Pt: 54…56 °C Flash Pt: None Storage Temperature: Ambient |
MDL Number:
MFCD00054657 CAS Number: 2062-98-8 EINECS: 218-173-8 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...