Diethyl tetrafluorosuccinate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16469-03
L16469-06
CAAAAL16469-03EA
44.56
CAD
CAAAAL16469-03
CAAAAL16469-06
Diethyl tetrafluorosuccinate 97%
Diethyl tetrafluorosuccinate
Purity: 97%
CAS No.: 377-71-9
Molecular Formula: C8H10F4O4
Formula Weight: 246.16
Refractive Index: 1.3680
Boiling Point: 78°/5mm
Beilstein Reference: 1798175
MDL No.: MFCD00015155
Formula:
C₈H₁₀F₄O₄ Boiling Pt: 78 °C/5 mm Density: 1.273 Storage Temperature: Ambient |
MDL Number:
MFCD00015155 CAS Number: 377-71-9 EINECS: 206-821-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...